missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5102925
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total CDC6.
The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.
Spécification
| Cdc6 | |
| Polyclonal | |
| Unconjugated | |
| CDC6 | |
| CDC18(S.pombe); CDC18L; cdc18-related protein; CDC6; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6; cell division cycle 6 homolog; HsCDC18; HsCDC6; p62(cdc6) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23834, 990 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O89033, Q99741 | |
| CDC6 | |
| A synthesized peptide derived from human CDC6(Accession Q99741), corresponding to amino acid residues L34-G84. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu