Learn More
Invitrogen™ Cdc5L Recombinant Superclonal™ Antibody (22HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711866
Description
This antibody is predicted to react with Monkey, Rabbit, Cat, Horse. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
The protein encoded by this gene shares a significant similarity with Schizosaccharomyces pombe cdc5 gene product, which is a cell cycle regulator important for G2/M transition. This protein has been demonstrated to act as a positive regulator of cell cycle G2/M progression. It was also found to be an essential component of a non-snRNA spliceosome, which contains at least five additional protein factors and is required for the second catalytic step of pre-mRNA splicing.
Specifications
| Cdc5L | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6A068, Q99459 | |
| CDC5L | |
| Protein corresponding to human Cdc5L [aa1-aa200]. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 22HCLC | |
| Unconjugated | |
| CDC5L | |
| 1200002I02Rik; AA408004; CDC5; CDC5 cell division cycle 5-like; CDC5 cell division cycle 5-like (S. pombe); CDC5L; CDC5-LIKE; Cdc5-like protein; Cdc5-related protein; CEF1; cell division cycle 5 like; cell division cycle 5-like; cell division cycle 5-like (S. pombe); cell division cycle 5-like protein; cell division cycle 5-related protein; dJ319D22.1; dJ319D22.1 (CDC5-like protein); hCDC5; KIAA0432; Pcdc5rp; Pombe cdc5-related protein; Unknown (protein for MGC:128482); zgc:55853 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 71702, 988 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.