Learn More
CDC5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12974-1-AP-20UL
 View more versions of this product
Product Code. 16810964
  • £135.00 / 20µL
Description
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.
Specifications
Specifications
CDC5L
Polyclonal
Unconjugated
CDC5L
Cdc5 like protein, CDC5L, CEF1, hCDC5, KIAA0432, PCDC5RP, Pombe cdc5 related protein
Rabbit
Antigen Affinity Chromatography
RUO
71702, 85434, 988
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08837, Q6A068, Q99459
Cdc5l
CDC5L Fusion Protein Ag3578
20 μL
Primary
Mouse, Rat, Human