missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDC5L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29317
This item is not returnable.
View return policy
Description
CDC5L Polyclonal antibody specifically detects CDC5L in Human samples. It is validated for Immunohistochemistry
Specifications
| CDC5L | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 988 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| CDC5, CDC5 (cell division cycle 5, S. pombe, homolog)-like, CDC5 cell division cycle 5-like (S. pombe), Cdc5-like protein, Cdc5-related protein, CEF1, cell division cycle 5-like protein, dJ319D22.1, dJ319D22.1 (CDC5-like protein), hCDC5, KIAA0432, PCDC5RPCDC5-LIKE, Pombe cdc5-related protein | |
| Between 752 and 802 | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction