missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC5L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-87013
This item is not returnable.
View return policy
Description
CDC5L Polyclonal specifically detects CDC5L in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| CDC5L | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| CDC5, CDC5 (cell division cycle 5, S. pombe, homolog)-like, CDC5 cell division cycle 5-like (S. pombe), Cdc5-like protein, Cdc5-related protein, CEF1, cell division cycle 5-like protein, dJ319D22.1, dJ319D22.1 (CDC5-like protein), hCDC5, KIAA0432, PCDC5RPCDC5-LIKE, Pombe cdc5-related protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 988 | |
| Human, Rat | |
| IgG |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2- 5 ug/mg lysate | |
| NP_001244.1 | |
| CDC5L | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human cell division cycle 5 (CDC5) -like using the numbering given in entry NP_001244.1 (GeneID988). | |
| 100 μL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with D. melanogaster, X. laevis, X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee and White spot disease agent_857967. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction