missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc45L Recombinant Rabbit Monoclonal Antibody (24GB3335)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557004
This item is not returnable.
View return policy
Description
Cdc45L Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene was identified by its strong similarity with Saccharomyces cerevisiae Cdc45, an essential protein required to the initiation of DNA replication. Cdc45 is a member of the highly conserved multiprotein complex including Cdc6/Cdc18, the minichromosome maintenance proteins (MCMs) and DNA polymerase, which is important for early steps of DNA replication in eukaryotes. This protein has been shown to interact with MCM7 and DNA polymerase alpha. Studies of the similar gene in Xenopus suggested that this protein play a pivotal role in the loading of DNA polymerase alpha onto chromatin. Multiple polyadenylation sites of this gene are reported.
Specifications
| Cdc45L | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cdc45 | |
| CDC 45; CDC 45L; Cdc45; CDC45 cell division cycle 45 homolog; CDC45 like; CDC45L; CDC45L2; CDC45-related protein; Cell division control protein 45 homolog; cell division cycle 45; cell division cycle 45 homolog; cell division cycle 45-like 2; human CDC45; PORC PI 1; PORC-PI-1; UNQ374/PRO710 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287961, 8318 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3335 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75419 | |
| Cdc45 | |
| A synthesized peptide derived from human CDC45 (aa 500-566). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction