missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc45L Monoclonal Antibody (OTI4F3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527419
This item is not returnable.
View return policy
Description
Cdc45L Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene was identified by its strong similarity with Saccharomyces cerevisiae Cdc45, an essential protein required to the initiation of DNA replication. Cdc45 is a member of the highly conserved multiprotein complex including Cdc6/Cdc18, the minichromosome maintenance proteins (MCMs) and DNA polymerase, which is important for early steps of DNA replication in eukaryotes. This protein has been shown to interact with MCM7 and DNA polymerase alpha. Studies of the similar gene in Xenopus suggested that this protein play a pivotal role in the loading of DNA polymerase alpha onto chromatin. Multiple polyadenylation sites of this gene are reported.
Specifications
| Cdc45L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75419 | |
| Cdc45 | |
| Human recombinant protein fragment corresponding to amino acids 1-270 of CDC45 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F3 | |
| Unconjugated | |
| Cdc45 | |
| CDC 45; CDC 45L; Cdc45; CDC45 cell division cycle 45 homolog; CDC45 like; CDC45L; CDC45L2; CDC45-related protein; Cell division control protein 45 homolog; cell division cycle 45; cell division cycle 45 homolog; cell division cycle 45-like 2; human CDC45; PORC PI 1; PORC-PI-1; UNQ374/PRO710 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8318 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction