missing translation for 'onlineSavingsMsg'
Learn More

CDC45L Antibody (3F11-1F3), Novus Biologicals™

Product Code. 18390309 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390309 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390309 Supplier Novus Biologicals Supplier No. H00008318M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDC45L Monoclonal antibody specifically detects CDC45L in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC45L
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 3F11-1F3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06232
Gene Alias CDC45 cell division cycle 45-like (S. cerevisiae), cell division cycle 45 homolog (S. cerevisiae), cell division cycle 45-like 2, human CDC45, S.cerevisiae, homolog)-like
Host Species Mouse
Immunogen CDC45L (AAH06232, 1 a.a. ~ 566 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MFVSDFRKEFYEVVQSQRVLLFVASDVDALCACKILQALFQCDHVQYTLVPVSGWQELETAFLEHKEQFHYFILINCGANVDLLDILQPDEDTIFFVCDTHRPVNVVNVYNDTQIKLLIKQDDDLEVPAYEDIFRDEEEDEEHSGNDSDGSEPSEKRTRLEEEIVEQTMRRRQRREWEARRRDILFDYEQYEYHGTSSAMVMFELAWMLSKDLNDMLWWAIVGLTDQWVQDKITQMKYVTDVGVLQRHVSRHNHRNEDEENTLSVDCTRISFEYDLRLVLYQHWSLHDSLCNTSYTAARFKLWSVHGQKRLQEFLADMGLPLKQVKQKFQAMDISLKENLREMIEESANKFGMKDMRVQTFSIHFGFKHKFLASDVVFATMSLMESPEKDGSGTDHFIQALDSLSRSNLDKLYHGLELAKKQLRATQQTIASCLCTNLVISQGPFLYCSLMEGTPDVMLFSRPASLSLLSKHLLKSFVCSTKNRRCKLLPLVMAAPLSMEHGTVTVVGIPPETDSSDRKNFFGRAFEKAAESTSSRMLHNHFDLSVIELKAEDRSKFLDALISLLS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8318
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.