missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc42EP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539085
This item is not returnable.
View return policy
Description
Cell division control protein 42 (CDC42), a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg (binder of Rho GTPases) family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to CDC42 and regulate its function negatively. The encoded protein may inhibit c-Jun N-terminal kinase (JNK) independently of CDC42 binding. The protein may also play a role in septin organization and inducing pseudopodia formation in fibroblasts [provided by RefSeq, Jul 2013].
Specifications
| Cdc42EP5 | |
| Polyclonal | |
| Unconjugated | |
| CDC42EP5 | |
| BORG3, CEP5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 148170, 58804 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6NZY7, Q9Z0X0 | |
| CDC42EP5 | |
| A synthetic peptide derived from the N-terminal region of human BORG3 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction