missing translation for 'onlineSavingsMsg'
Learn More

CDC42EP2 Antibody, HRP, Novus Biologicals™

Product Code. 18322412
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322412 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322412 Supplier Bio-Techne Supplier No. NBP306534H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC42EP2 Polyclonal antibody specifically detects CDC42EP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42EP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias BORG1CEP2Binder of Rho GTPases 1, CDC42 effector protein (Rho GTPase binding) 2, cdc42 effector protein 2, CRIB-containing BOGR1 protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CDC42EP2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10435
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.