missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc42EP1 Recombinant Rabbit Monoclonal Antibody (23GB5745), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Cdc42EP1 |
|---|---|
| Clone | 23GB5745 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087565
![]() |
Invitrogen™
MA552904 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087877
|
Invitrogen™
MA552905 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdc42EP1 Recombinant Monoclonal antibody specifically detects Cdc42EP1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Cdc42EP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q00587 | |
| 11135 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5745 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1810058K22Rik; 55 kDa bone marrow stromal/endothelial cell protein; AA980734; binder of Rho GTPase 5; binder of Rho GTPases 5; Borg5; CDC42 effector protein (Rho GTPase binding) 1; cdc42 effector protein 1; Cdc42ep1; cdc42ep1 {ECO:0000312; CEP1; EMBL:AAI28745.1}; MSE55; serum constituent protein; serum protein MSE55 | |
| A synthesized peptide derived from human CDC42EP1 (300-391AA). | |
| Primary | |
| -20°C | |
| CDC42EP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title