missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42EP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17693-100UL
This item is not returnable.
View return policy
Beskrivning
CDC42EP1 Polyclonal antibody specifically detects CDC42EP1 in Human samples. It is validated for Immunofluorescence
Specifikationer
| CDC42EP1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Binder of Rho GTPases 5, Borg5, BORG5cdc42 effector protein 1, CDC42 effector protein (Rho GTPase binding) 1, CEP155 kDa bone marrow stromal/endothelial cell protein, MSE55MGC15316, serum constituent protein, Serum protein MSE55 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GAGWDGGHHYPEMDARQERVEVLPQARASWESLDEEWRAPQAGSRTPVPSTVQANTFEFADAEEDDEVKV | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 11135 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering