missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc42EP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103722
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BORG5.
CDC42 is a member of the Rho GTPase family that regulates multiple cellular activities, including actin polymerization. The protein encoded by this gene is a CDC42 binding protein that mediates actin cytoskeleton reorganization at the plasma membrane. This protein is secreted and is primarily found in bone marrow.
Specifications
| Cdc42EP1 | |
| Polyclonal | |
| Unconjugated | |
| CDC42EP1 | |
| 1810058K22Rik; 55 kDa bone marrow stromal/endothelial cell protein; AA980734; binder of Rho GTPase 5; binder of Rho GTPases 5; Borg5; CDC42 effector protein (Rho GTPase binding) 1; cdc42 effector protein 1; Cdc42ep1; cdc42ep1 {ECO:0000312; CEP1; EMBL:AAI28745.1}; MSE55; serum constituent protein; serum protein MSE55 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 104445, 11135 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q00587, Q91W92 | |
| CDC42EP1 | |
| A synthesized peptide derived from human CDC42EP1(Accession Q00587), corresponding to amino acid residues R53-P103. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction