missing translation for 'onlineSavingsMsg'
Learn More

CDC42EP1 Antibody (3A4), Novus Biologicals™

Product Code. 18379329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379329 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379329 Supplier Novus Biologicals Supplier No. H00011135M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDC42EP1 Monoclonal antibody specifically detects CDC42EP1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42EP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3A4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH09356
Gene Alias Binder of Rho GTPases 5, Borg5, BORG5cdc42 effector protein 1, CDC42 effector protein (Rho GTPase binding) 1, CEP155 kDa bone marrow stromal/endothelial cell protein, MSE55MGC15316, serum constituent protein, Serum protein MSE55
Host Species Mouse
Immunogen CDC42EP1 (AAH09356, 1 a.a. ~ 384 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPGPQGGRGAATMSLGKLSPVGWVSSSQGKRRLTADMISHPLGDFRHTMHVGRGGDVFGDTSFLSNHGGSSGSTHRSPRSFLAKKLQLVRRVGAPPRRMASPPAPSPAPPAISPIIKNAISLPQLNQAAYDSLVVGKLSFDSSPTSSTDGHSSYGLDSGFCTISRLPRSEKPHDRDRDGSFPSEPGLRRSDSLLSFRLDLDLGPSLLSELLGVMSLPEAPAAETPAPAANPPAPTANPTGPAANPPATTANPPAPAATPTGPAANPPAPAASSTPHGHCPNGVTAGLGPVAEVKSSPVGGGPRGPAGPALGRHWGAGWDGGHHYPEMDARQERVEVLPQARASWESLDEEWRAPQAGSRTPVPSTVQANTFEFADAEEDDEVKV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11135
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.