missing translation for 'onlineSavingsMsg'
Learn More

CDC42BPG Antibody, Novus Biologicals™

Product Code. 18334509 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334509 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334509 Supplier Novus Biologicals Supplier No. H00055561A01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CDC42BPG Polyclonal antibody specifically detects CDC42BPG in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42BPG
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation 50% Glycerol
Gene Accession No. NP_059995
Gene Alias CDC42 binding protein kinase gamma (DMPK-like), CDC42-binding protein kinase gamma, DMPK2MRCK gamma, DMPK-like gamma, EC 2.7.11, EC 2.7.11.1, HSMDPKIN, kappa-200, MRCKgamma, Myotonic dystrophy kinase-related CDC42-binding kinase gamma, myotonic dystrophy protein kinase like protein, Myotonic dystrophy protein kinase-like alpha, Myotonic dystrophy protein kinase-like gamma, serine/threonine-protein kinase MRCK gamma
Host Species Mouse
Immunogen CDC42BPG (NP_059995, 1423 a.a. - 1528 a.a.) partial recombinant protein with GST tag. RREMLKDPFVRSKLISPPTNFNHLVHVGPANGRPGARDKSPAPEEKGRVARGSGPQRPHSFSEALRRPASMGSEGLGGDADPMKRKPWTSLSSESVSCPQGSLSPA
Purification Method Protein A purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55561
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.