missing translation for 'onlineSavingsMsg'
Learn More

CDC42BPB Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18322845 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322845 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322845 Supplier Novus Biologicals Supplier No. NBP299815AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC42BPB Polyclonal specifically detects CDC42BPB in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42BPB
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias CDC42 binding protein kinase beta (DMPK-like), CDC42-binding protein kinase beta, CDC42-binding protein kinase beta (DMPK-like), DMPK-like beta, EC 2.7.11, EC 2.7.11.1, FLJ37601, FLJ44730, KIAA1124DKFZp686H1431, MRCK beta, MRCKB, Myotonic dystrophy kinase-related CDC42-binding kinase beta, Myotonic dystrophy protein kinase-like beta, serine/threonine-protein kinase MRCK beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CDC42BPB (Q9Y5S2; Met1-His427).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9578
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.