missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc42 Recombinant Rabbit Monoclonal Antibody (23GB1025), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553949
This item is not returnable.
View return policy
Description
Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.
Specifications
| Cdc42 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CDC42 | |
| AI747189; AU018915; CDC42; CDC42Hs; Cell division control protein 42 homolog; cell division cycle 42; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K; G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; GTP-binding protein; RP1-224A6.5; small GTP binding protein CDC42; TKS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12540, 64465, 998 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1025 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60766, P60953, Q8CFN2 | |
| CDC42 | |
| Recombinant protein of human CDC42 (60-191AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction