missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc42 Polyclonal Antibody

Description
Cdc42 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for the Rac1 gene.
Especificaciones
Especificaciones
| Antigen | Cdc42 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide |
| Gene | CDC42 |
| Gene Accession No. | P60766, P60953, Q8CFN2 |
| Gene Alias | AI747189; AU018915; CDC42; CDC42Hs; Cell division control protein 42 homolog; cell division cycle 42; cell division cycle 42 (GTP binding protein); cell division cycle 42 homolog; dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)); dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)); G25K; G25K GTP-binding protein; growth-regulating protein; GTP binding protein, 25kDa; GTP-binding protein; RP1-224A6.5; small GTP binding protein CDC42; TKS |
| Gene Symbols | CDC42 |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Cdc42 Polyclonal Antibody
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: sodium azide
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?