missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
£150.00
Specifications
| Antigen | CDC42 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CDC42 Polyclonal specifically detects CDC42 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| CDC42 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Chicken | |
| Breast Cancer, Cancer, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42 | |
| CDC42 | |
| IgY | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_001782 | |
| 998 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human CDC42. The immunogen is located within amino acids 40 - 90 of CDC42. Amino Acid Squence: FDTAGQEDYDRLRPLSY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title