missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc40 Recombinant Rabbit Monoclonal Antibody (2I8I7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538004
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, HCT116, HepG2, Mouse lung, Mouse brain, Mouse spleen, Rat lung Immunogen sequence: MSAAIAALAA SYGSGSGSES DSDSESSRCP LPAADSLMHL TKSPSSKPSL AVAVDSAPEV AVKEDLETGV HLDPAVKEVQ YNPTYETMFA PEFGPENPFR.
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is found to be essential for the catalytic step II in pre-mRNA splicing process. It is found in the spliceosome, and contains seven WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp17 protein, which functions in two different cellular processes: pre-mRNA splicing and cell cycle progression. It suggests that this protein may play a role in cell cycle progression.
Specifications
| Cdc40 | |
| Recombinant Monoclonal | |
| 0.28 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60508, Q9DC48 | |
| CDC40 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDC40 (O60508). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2I8I7 | |
| Unconjugated | |
| CDC40 | |
| 1200003H23Rik; Cdc40; cell division cycle 40; cell division cycle 40 homolog; EHB3; EH-binding protein 3; hPRP17; pre-mRNA splicing factor 17; pre-mRNA-processing factor 17; PRP17; PRP17 homolog; PRPF17 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 361859, 51362, 71713 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction