missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cdc40 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15968194
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15968194 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15968194 Supplier Invitrogen™ Supplier No. PA565451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VQYNPTYETMF APEFGPENPF RTQQMAAPRN MLSGYAEPAH INDFMFEQQR RTFATYGYAL DPSLDNHQVS AKYIGSVEEA EKNQGLTVFE TGQKKT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 73%.

Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is found to be essential for the catalytic step II in pre-mRNA splicing process. It is found in the spliceosome, and contains seven WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp17 protein, which functions in two different cellular processes: pre-mRNA splicing and cell cycle progression. It suggests that this protein may play a role in cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc40
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC40
Gene Accession No. O60508
Gene Alias 1200003H23Rik; Cdc40; cell division cycle 40; cell division cycle 40 homolog; EHB3; EH-binding protein 3; hPRP17; pre-mRNA splicing factor 17; pre-mRNA-processing factor 17; PRP17; PRP17 homolog; PRPF17
Gene Symbols CDC40
Host Species Rabbit
Immunogen Recombinant Human Cdc40. Recombinant protein control fragment (Product #RP-106131).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51362
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.