missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDC40 Antibody (SR3222), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | CDC40 |
|---|---|
| Clone | SR3222 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041683
|
Novus Biologicals™
NBP3-27760-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041805
|
Novus Biologicals™
NBP3-27760-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC40 Monoclonal antibody specifically detects CDC40 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| CDC40 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cell division cycle 40 homolog, cell division cycle 40 homolog (S. cerevisiae), cell division cycle 40 homolog (yeast), Ehb3, EH-binding protein 3, hPRP17, MGC102802, pre-mRNA splicing factor 17, pre-mRNA-processing factor 17, PRP17 homolog, PRP17EHB3PRPF17FLJ10564 | |
| A synthesized peptide derived from human CDC40 (Uniprot #: O60508) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3222 | |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 51362 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title