missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC40 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12781APC
This item is not returnable.
View return policy
Description
CDC40 Polyclonal antibody specifically detects CDC40 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CDC40 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| Cell division cycle 40 homolog, cell division cycle 40 homolog (S. cerevisiae), cell division cycle 40 homolog (yeast), Ehb3, EH-binding protein 3, hPRP17, MGC102802, pre-mRNA splicing factor 17, pre-mRNA-processing factor 17, PRP17 homolog, PRP17EHB3PRPF17FLJ10564 | |
| The immunogen this antibody was made to, maps to a region between residue 175 and 225 of human Cell Division Cycle 40 using the numbering given in entry NP_056975.1 (GeneID 51362). | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 51362 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction