missing translation for 'onlineSavingsMsg'
Learn More

CDC40 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18424586 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424586 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424586 Supplier Novus Biologicals Supplier No. NBP212781AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC40 Polyclonal antibody specifically detects CDC40 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC40
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Cell division cycle 40 homolog, cell division cycle 40 homolog (S. cerevisiae), cell division cycle 40 homolog (yeast), Ehb3, EH-binding protein 3, hPRP17, MGC102802, pre-mRNA splicing factor 17, pre-mRNA-processing factor 17, PRP17 homolog, PRP17EHB3PRPF17FLJ10564
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 175 and 225 of human Cell Division Cycle 40 using the numbering given in entry NP_056975.1 (GeneID 51362).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51362
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.