missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc37L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521853
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM, Mar 2008].
Specifications
| Cdc37L1 | |
| Polyclonal | |
| Unconjugated | |
| CDC37L1 | |
| 2700033A15Rik; BB159850; CDC37 cell division cycle 37 homolog-like 1; CDC37B; Cdc37l; Cdc37l1; cell division cycle 37 homolog-like 1; cell division cycle 37 like 1; cell division cycle 37-like 1; HARC; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37; RP11-6J24.5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55664 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.59 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q7L3B6 | |
| CDC37L1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CDC37L1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction