missing translation for 'onlineSavingsMsg'
Learn More

CDC37L1 Antibody (1D4), Novus Biologicals™

Product Code. 18325199 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325199 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325199 Supplier Novus Biologicals Supplier No. H00055664M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDC37L1 Monoclonal antibody specifically detects CDC37L1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC37L1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14133
Gene Alias CDC37 cell division cycle 37 homolog (S. cerevisiae)-like 1, CDC37BHARCCDC37 cell division cycle 37 homolog-like 1, cell division cycle 37 homolog (S. cerevisiae)-like 1, FLJ20639, hsp90 co-chaperone Cdc37-like 1, Hsp90-associating relative of Cdc37
Host Species Mouse
Immunogen CDC37L1 (AAH14133, 1 a.a. ~ 337 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEQPWPPPGPWSLPRAEGEAEEESDFDVFPSSPRCPQLPGGGAQMYSHGIELACQKQKEFVKSSVACKWNLAEAQQKLGSLALHNSESLDQEHAKAQTAVSELRQREEEWRQKEEALVQREKMCLWSTDAISKDVFNKSFINQDKRKDTEDEDKSESFMQKYEQKIRHFGMLSRWDDSQRFLSDHPYLVCEETAKYLILWCFHLEAEKKGALMEQIAHQAVVMQFIMEMAKNCNVDPRGCFRLFFQKAKAEEEGYFEAFKNELEAFKSRVRLYSQSQSFQPMTVQNHVPHSGVGSIGLLESLPQNPDYLQYSISTALCSLNSVVHKEDDEPKMMDTV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55664
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.