missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37, p Ser13 Antibody (3B3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26429-50ul
This item is not returnable.
View return policy
Description
CDC37 Monoclonal antibody specifically detects CDC37 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CDC37 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3B3 | |
| Western Blot 1:500-1:5000, ELISA | |
| CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
| A synthesized peptide derived from Human CDC37 [p Ser13] [UniProt Q16543] | |
| 50 μL | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific | |
| 11140 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction