missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37 Mouse anti-Human, Mouse, Rat, Clone: 1C2A6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CDC37 |
|---|---|
| Clone | 1C2A6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818285
|
Proteintech
66420-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808285
|
Proteintech
66420-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In S. cerevisiae, Cdc37 has been shown to be an important regulator of cell division cycle through its action on cyclin-dependent kinases (CDK′s). Cdc37 has also been associated with signaling pathways and kinases unrelated to cell cycle control, including v-Src, casein kinase II, MPS1 kinase and the sevenless receptor tyrosine kinase. Interestingly, HSP90 has also been shown to be associated with many of the same cellular targets as Cdc37. It has now been shown that Cdc37 is the same protein previously described as p50 which has been found to be associated with many of the same proteins as HSP90. Among others, Cdc37/p50 has been found complexed with Src-family kinases (Fyn, Yes, Fes, Lck & Fgr), Raf and pp60v-src. More recently, Cdc37 has been shown to be a chaperone independently and in concert with HSP90. In fact, Cdc37 is necessary for the binding of HSP90 to Cdk4.Specifications
| CDC37 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CDC37, CDC37A, Hsp90 co chaperone Cdc37, P50CDC37 | |
| Cdc37 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1C2A6 | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Rat, Human, Mouse | |
| Q16543, Q61081, Q63692 | |
| 11140, 114562, 12539 | |
| CDC37 Fusion Protein Ag0267 | |
| Primary | |
| -20°C | |
| CDC37 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title