missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37 Mouse anti-Human, Mouse, Clone:6B3B7, Abnova™
Mouse monoclonal antibody raised against recombinant human CDC37.
Brand: Abnova MAB17279.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is highly similar to Cdc 37, a cell division cycle control protein of Sacchromyces cerevisiae. This protein is a molecular chaperone with specific function in cell signal transduction. It has been shown to form complex with Hsp90 and a variety of protein kinases including CDK4, CDK6, SRC, RAF-1, MOK, as well as eIF2 alpha kinases. It is thought to play a critical role in directing Hsp90 to its target kinases. [provided by RefSeq]Specifications
| CDC37 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| CDC37 | |
| Mouse | |
| 100 μg | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Western Blot | |
| 6B3B7 | |
| cell division cycle 37 homolog (S. cerevisiae) | |
| In PBS (0.05% sodium azide) | |
| P50CDC37 | |
| Recombinant protein corresponding to amino acid 241-378 of human CDC37 from E. coli. | |
| 11140 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction