missing translation for 'onlineSavingsMsg'
Learn More

CDC37 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18330362 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330362 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330362 Supplier Novus Biologicals Supplier No. NBP298986G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC37 Polyclonal specifically detects CDC37 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC37
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human CDC37.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 11140
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.