missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc34 Recombinant Rabbit Monoclonal Antibody (23GB3435), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Cdc34 |
|---|---|
| Clone | 23GB3435 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089955
![]() |
Invitrogen™
MA552608 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088644
|
Invitrogen™
MA552609 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdc34 Recombinant Monoclonal antibody specifically detects Cdc34 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Cdc34 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P49427, Q8CFI2 | |
| 216150, 299602, 997 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3435 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| (E3-independent) E2 ubiquitin-conjugating enzyme R1; AI327276; CDC34; cell division cycle 34; cell division cycle 34 homolog; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; serine protease; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme Cdc34; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
| A synthetic peptide of human CDC34 (2-236AA). | |
| Primary | |
| -20°C | |
| CDC34 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title