missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc2L6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599764
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CD2L6.
CDC2L6 is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.
Specifications
| Cdc2L6 | |
| Polyclonal | |
| Unconjugated | |
| Cdk19 | |
| 2700084L06Rik; AI845279; AW228747; bA346C16.3; Cdc2l6; CDC2-related protein kinase 6; Cdk11; Cdk19; CDK8-like cyclin-dependent kinase; cell division cycle 2-like 6 (CDK8-like); cell division cycle 2-like protein kinase 6; Cell division protein kinase 19; cyclin dependent kinase 19; cyclin dependent kinase 19 variant 2; cyclin-dependent kinase (CDC2-like) 11; cyclin-dependent kinase 11; Cyclin-dependent kinase 19; death-preventing kinase; KIAA1028; mKIAA1028 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23097, 78334 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BWD8, Q9BWU1 | |
| Cdk19 | |
| A synthesized peptide derived from human CDK19(Accession Q9BWU1), corresponding to amino acid residues L431-L481. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction