missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDC2L6 Antibody (CL14114), Novus Biologicals™
Mouse Monoclonal Antibody
£336.00 - £483.00
Specifications
| Antigen | CDC2L6 |
|---|---|
| Clone | CL14114 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043549
|
Novus Biologicals™
NBP3-26849-100ul |
100 μL |
£483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043738
|
Novus Biologicals™
NBP3-26849-25ul |
25 μL |
£336.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC2L6 Monoclonal antibody specifically detects CDC2L6 in Human samples. It is validated for Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| CDC2L6 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| PBS, pH 7.2, 40% glycerol | |
| 23097 | |
| IgG2b | |
| Protein A purified |
| CL14114 | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bA346C16.3, CDC2L6, CDC2-related protein kinase 6, CDK8-like cyclin-dependent kinase, cell division cycle 2-like 6 (CDK8-like), Cell division cycle 2-like protein kinase 6, Cell division protein kinase 19, cyclin-dependent kinase (CDC2-like) 11, Cyclin-dependent kinase 11, cyclin-dependent kinase 19, Death-preventing kinase, EC 2.7.11, EC 2.7.11.22, KIAA1028CDK11 | |
| This antibody was developed against a Synthetic Peptide of Q9BWU1. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title