Learn More
CDC2L6 Antibody (CL14114) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26849UV
Product Code. 30291469
  • £599.00 / 0.10mL
Description
CDC2L6 Monoclonal antibody specifically detects CDC2L6 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDC2L6
Monoclonal
DyLight 350
bA346C16.3, CDC2L6, CDC2-related protein kinase 6, CDK8-like cyclin-dependent kinase, cell division cycle 2-like 6 (CDK8-like), Cell division cycle 2-like protein kinase 6, Cell division protein kinase 19, cyclin-dependent kinase (CDC2-like) 11, Cyclin-dependent kinase 11, cyclin-dependent kinase 19, Death-preventing kinase, EC 2.7.11, EC 2.7.11.22, KIAA1028CDK11
This antibody was developed using a synthetic peptide derived from Q9BWU1, with the exact immunogen sequence remaining proprietary.
0.1 mL
Protein Kinase
23097
Store at 4⪚C in the dark.
IgG2b
Western Blot, Immunocytochemistry/Immunofluorescence
CL14114
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified