missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDC2L5 Polyclonal specifically detects CDC2L5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | CDC2L5 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q14004 |
| Gene Alias | CDC2L5, CDC2LEC 2.7.11.22, CDC2-related protein kinase 5, Cell division cycle 2-like protein kinase 5, Cell division protein kinase 13, CHEDcell division cycle 2-like 5 (cholinesterase-related cell division controller), Cholinesterase-related cell division controller, cyclin-dependent kinase 13, EC 2.7.11, EC 2.7.11.23, hCDK13, KIAA1791FLJ35215 |
| Gene Symbols | CDK13 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?