missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDC27 Recombinant Mouse Monoclonal Antibody (K70013_5G6)

Product Code. 30828588
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828588 100 μL
30825885 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828588 Supplier Invitrogen™ Supplier No. MA564428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: TLNKAIVIDP KNPLCKFHRA SVLFANEKYK SALQELEELK QIVPKESLVY FLIGKVYKKL GQTHLALMNF SWAMDLDPKG ANNQIKEAID KRYLPDDEEP ITQEEQIMGT DESQESSMTD ADDTQLHAAE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%, Pig - 100%, Cynomolgus monkey - 100%.

The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae protein Cdc27, and the gene product of Schizosaccharomyces pombe nuc 2. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction. This protein was shown to interact with mitotic checkpoint proteins including Mad2, p55CDC and BUBR1, and thus may be involved in controlling the timing of mitosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC27
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K70013_5G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CDC27
Gene Accession No. P30260
Gene Alias AI452358; AN CDC27Hs; ANAPC3; anaphase promoting complex subunit 3; Anaphase-promoting complex subunit 3; anaphase-promoting complex, protein 3; APC3; BC023187; cdc27; CDC27 homolog; cdc27.S; CDC27Hs; cell division cycle 27; cell division cycle 27 homolog; cell division cycle 27 homolog (S. cerevisiae); cell division cycle 27 S homeolog; cell division cycle protein 27 homolog; D0S1430E; D17S978E; HNUC; H-NUC; hypothetical protein; NUC2; nuc2 homolog; XELAEV_18046079mg
Gene Symbols CDC27
Host Species Mouse
Immunogen Recombinant human CDC27 (aa 691-820).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 996
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.