Learn More
CDC27 APC3 Mouse anti-Human, Unconjugated, Clone: 2F10G2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CDC27 APC3 |
|---|---|
| Clone | 2F10G2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889755
|
Proteintech
67239-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879755
|
Proteintech
67239-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CDC27; APC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC27; APC3. This antibody recognizes human, mouse, rat antigen. The CDC27; APC3 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
| CDC27 APC3 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 996 | |
| Primary |
| 2F10G2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.