missing translation for 'onlineSavingsMsg'
Learn More

Cdc27 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18406027 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406027 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406027 Supplier Novus Biologicals Supplier No. NBP177377MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cdc27 Polyclonal antibody specifically detects Cdc27 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc27
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human APC3. The immunogen is located within the last 50 amino acids of APC3. Amino Acid Squence: EAIDKRYLPDDEEPITQEE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 996
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.