missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc27 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77377-0.025mg
This item is not returnable.
View return policy
Description
Cdc27 Polyclonal specifically detects Cdc27 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cdc27 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_001247 | |
| CDC27 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human APC3. The immunogen is located within the last 50 amino acids of APC3. Amino Acid Squence: EAIDKRYLPDDEEPITQEE | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 996 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction