missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100434
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CDC26.
CDC26, also named as ANAPC12, APC12 and C9orf17, is a component of the anaphase promoting complex/cyclosome (APC/C).
Specifications
| Cdc26 | |
| Polyclonal | |
| Unconjugated | |
| CDC26 | |
| 2010012C09Rik; ANAPC12; anaphase-promoting complex subunit 12; Anaphase-promoting complex subunit CDC26; APC12; BWK-2; C9orf17; CDC26; CDC26 subunit of anaphase promoting complex; cell division cycle 26; cell division cycle 26 homolog; cell division cycle protein 26 homolog; Protein BWK-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 246184, 366381, 66440 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6YDN7, Q8NHZ8, Q99JP4 | |
| CDC26 | |
| A synthesized peptide derived from human CDC26(Accession Q8NHZ8), corresponding to amino acid residues E37-F85. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction