missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDC25B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104215
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CDC25B.
Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.
Specifications
| CDC25B | |
| Polyclonal | |
| Unconjugated | |
| CDC25B | |
| AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12531, 171103, 994 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30305, P30306, P48966 | |
| CDC25B | |
| A synthesized peptide derived from human CDC25B(Accession P30305), corresponding to amino acid residues V298-N348. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction