missing translation for 'onlineSavingsMsg'
Learn More

CDC25A Mouse anti-Human, Clone: DCS121, Novus Biologicals™

Product Code. 18308462 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18308462 25 μg 25µg
18390103 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18308462 Supplier Novus Biologicals Supplier No. NBP31188225UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDC25A Monoclonal antibody specifically detects CDC25A in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC25A
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone DCS121
Conjugate Unconjugated
Dilution Western Blot 1 - 10 ug/ml, Immunoprecipitation 1 - 10 ug/ml
Formulation PBS
Gene Alias CDC25A2, CDC25A2-CAG isoform, cell division cycle 25 homolog A (S. cerevisiae), cell division cycle 25 homolog A (S. pombe), cell division cycle 25A, Dual specificity phosphatase Cdc25A, EC 3.1.3.48, M-phase inducer phosphatase 1
Host Species Mouse
Immunogen Mice were immunized with a purified recombinant CDC25A protein of 65kD protein (Uniprot: P30304)
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 993
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.