Learn More
Cdc25A Monoclonal Antibody (DCS-120), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA513794
Product Code. 11324923
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
MA5-13794 targets CDC25A in IP, IHC (P) and WB applications and shows reactivity with Human and Rat samples. The MA5-13794 immunogen is purified recombinant CDC25A.
CDC25a is a member of the CDC25 family of phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. CDC25a is required for progression through G1 and S phases of the cell cycle. CDC25a activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division. CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cdc25A
Monoclonal
0.2 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P30304, P48964, P48965
CDC25A
Purified recombinant CDC25A
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
DCS-120
Unconjugated
CDC25A
CDC25 isoform A1-CAG; CDC25A; CDC25A2; CDC25A2-CAG isoform; Cdc25m3; cell division cycle 25A; D9Ertd393e; dual specificity phosphatase Cdc25A; EC 3.1.3.48; M-phase inducer phosphatase 1; MPIP1
Mouse
Protein A
RUO
12530, 171102, 993
4° C