missing translation for 'onlineSavingsMsg'
Learn More

CDC25A Antibody (336445) [DyLight 350], Novus Biologicals™

Product Code. 30735120 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735120 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735120 Supplier Novus Biologicals Supplier No. FAB1648D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDC25A Monoclonal antibody specifically detects CDC25A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC25A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 336445
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CDC25A2, CDC25A2-CAG isoform, cell division cycle 25 homolog A (S. cerevisiae), cell division cycle 25 homolog A (S. pombe), cell division cycle 25A, Dual specificity phosphatase Cdc25A, EC 3.1.3.48, M-phase inducer phosphatase 1
Host Species Mouse
Immunogen E. coli-derived recombinant human CDC25A, Glu2-Leu523, Accession # AAA58415
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 993
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.