missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cdc23 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12809102
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809102 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809102 Supplier Invitrogen™ Supplier No. PA520949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-20949 can be used with blocking peptide PEP-1063.

Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). Ubr2 (Ubiquitin-protein ligase E3-alpha-2), also known as N-recognin-2, is a 1755 amino acid protein that contains one UBR-type zinc finger and one RING-type zinc finger. Participating in protein modification events within the N-end rule pathway, Ubr2 functions as an E3 ubiquitin-protein ligase that recognizes and binds proteins that contain destabilizing N-terminal residues, thereby leading to their ubiquitination and subsequent degradation. Mice lacking Ubr2 are infertile due to defects in male meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc23
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Cdc23
Gene Accession No. Q8BGZ4, Q9UJX2
Gene Alias 6030435O18; ANAPC8; anaphase promoting complex subunit 8; anaphase-promoting complex subunit 8; APC8; CDC23; CDC23 (cell division cycle 23, yeast, homolog); CDC23 cell division cycle 23; cell division cycle 23; cell division cycle 23 homolog; cell division cycle protein 23; cell division cycle protein 23 homolog; CUT23; cyclosome subunit 8; D18Ertd243e; zgc:56013
Gene Symbols Cdc23
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human APC8.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291689, 52563, 8697
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.