missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570427
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). Ubr2 (Ubiquitin-protein ligase E3-alpha-2), also known as N-recognin-2, is a 1755 amino acid protein that contains one UBR-type zinc finger and one RING-type zinc finger. Participating in protein modification events within the N-end rule pathway, Ubr2 functions as an E3 ubiquitin-protein ligase that recognizes and binds proteins that contain destabilizing N-terminal residues, thereby leading to their ubiquitination and subsequent degradation. Mice lacking Ubr2 are infertile due to defects in male meiosis.
Specifications
| Cdc23 | |
| Polyclonal | |
| Unconjugated | |
| Cdc23 | |
| 6030435O18; ANAPC8; anaphase promoting complex subunit 8; anaphase-promoting complex subunit 8; APC8; CDC23; CDC23 (cell division cycle 23, yeast, homolog); CDC23 cell division cycle 23; cell division cycle 23; cell division cycle 23 homolog; cell division cycle protein 23; cell division cycle protein 23 homolog; CUT23; cyclosome subunit 8; D18Ertd243e; zgc:56013 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 291689 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Cdc23 | |
| synthetic peptide directed towards the N-terminal region of rat Cdc23. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction