missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cdc23 Polyclonal antibody specifically detects Cdc23 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Cdc23 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ANAPC8anaphase promoting complex subunit 8, Anaphase-promoting complex subunit 8, APC8CDC23 (cell division cycle 23, yeast, homolog), cell division cycle 23 homolog (S. cerevisiae), cell division cycle protein 23 homolog, CUT23, Cyclosome subunit 8 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the center of human APC8. The immunogen is located within amino acids 270 - 320 of APC8. Amino Acid Squence: FNELRKQDPYRIENMDT |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?