missing translation for 'onlineSavingsMsg'
Learn More

Cdc23 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18417852 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417852 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417852 Supplier Novus Biologicals Supplier No. NBP177156AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cdc23 Polyclonal antibody specifically detects Cdc23 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc23
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ANAPC8anaphase promoting complex subunit 8, Anaphase-promoting complex subunit 8, APC8CDC23 (cell division cycle 23, yeast, homolog), cell division cycle 23 homolog (S. cerevisiae), cell division cycle protein 23 homolog, CUT23, Cyclosome subunit 8
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human APC8. The immunogen is located within amino acids 270 - 320 of APC8. Amino Acid Squence: FNELRKQDPYRIENMDT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8697
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.