missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cdc20 Recombinant Rabbit Monoclonal Antibody (3P9Y9)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557921

 View more versions of this product

Product Code. 30283769

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAQFAFESDL HSLLQLDAPI PNAPPARWQR KAKEAAGPAP SPMRAANRSH SAGRTPGRTP GKSSSKVQTT PSKPGGDRYI PHRSAAQMEV ASFLLSKENQ.

Required for full ubiquitin ligase activity of the anaphase promoting complex/cyclosome (APC/C) and may confer substrate specificity upon the complex. Is regulated by MAD2L1: in metaphase the MAD2L1-CDC20-APC/C ternary complex is inactive and in anaphase the CDC20-APC/C binary complex is active in degrading substrates. The CDC20-APC/C complex positively regulates the formation of synaptic vesicle clustering at active zone to the presynaptic membrane in postmitotic neurons. CDC20-APC/C-induced degradation of NEUROD2 induces presynaptic differentiation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cdc20
Recombinant Monoclonal
1.53 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q12834, Q62623
CDC20
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDC20 (NP_001246.2).
20 μL
Primary
Human, Rat
Antibody
IgG
ELISA, Immunoprecipitation, Western Blot
3P9Y9
Unconjugated
CDC20
2310042N09Rik; bA276H19.3; C87100; cdc20; CDC20 cell division cycle 20 homolog; cdc20.S; CDC20A; cell cycle protein p55CDC; cell division cycle 20; cell division cycle 20 homolog; cell division cycle 20 S homeolog; cell division cycle protein 20 homolog; fizzy1; mmCdc20; p55cdc; XELAEV_18025163mg; X-FZY
Rabbit
Protein A
RUO
64515, 991
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.