missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cdc20 Monoclonal specifically detects Cdc20 in Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
Specifications
| Antigen | Cdc20 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | BA8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, CyTOF-ready |
| Gene Alias | CDC20 cell division cycle 20 homolog, CDC20 cell division cycle 20 homolog (S. cerevisiae), cell division cycle 20 homolog (S. cerevisiae), cell division cycle protein 20 homolog, MGC102824, p55CDCS. cerevisiae, homolog) |
| Gene Symbols | CDC20 |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?